siRNA Details
| Gene Symbol: Rbm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148930.3 | NP_683732.1 | 4, 5 | 505 | ||
| GenBank mRNA | AJ309168.1 | - | 3, 4 | 340 | ||
| AK133999.1 | - | 4, 5 | 463 | |||
| AK134183.1 | - | 4, 5 | 500 | |||
| AK146880.1 | - | 4, 5 | 500 | |||
| BC019766.1 | - | 4, 5 | 450 | |||
| BC023854.1 | - | 4, 5 | 490 | 5346780 | ||
| BC031899.1 | - | 4, 5 | 490 | 5362121 | ||
| BC042419.1 | - | 4, 5 | 483 | |||
| BC043058.1 | - | 4, 5 | 442 | 6419695 | ||
| BC054729.1 | - | 4, 5 | 493 | 6419758 | ||
| BC060985.1 | - | 4, 5 | 483 | |||

Rbm5
Genomic Map