siRNA Details
Gene Symbol: Gsdmc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031378.3 | NP_113555.1 | 4 | 787 | ||
GenBank mRNA | AB042406.1 | - | 3 | 684 | ||
AK137662.1 | - | 4 | 788 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gsdmc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031378.3 | NP_113555.1 | 4 | 787 | ||
GenBank mRNA | AB042406.1 | - | 3 | 684 | ||
AK137662.1 | - | 4 | 788 |