siRNA Details
| Gene Symbol: Tex13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031381.2 | NP_113558.1 | 2 | 143 | ||
| GenBank mRNA | AF285576.1 | - | 2 | 143 | ||
| BC116863.1 | - | 2 | 119 | 40126182 | ||
| BC119125.1 | - | 2 | 119 | 8733874 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex13| Gene Symbol: Tex13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031381.2 | NP_113558.1 | 2 | 143 | ||
| GenBank mRNA | AF285576.1 | - | 2 | 143 | ||
| BC116863.1 | - | 2 | 119 | 40126182 | ||
| BC119125.1 | - | 2 | 119 | 8733874 | ||