siRNA Details
| Gene Symbol: Tdrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002238.2 | NP_001002238.1 | 23 | 3452 | ||
| NM_001002240.2 | NP_001002240.1 | 23 | 3425 | |||
| NM_001002241.2 | NP_001002241.1 | 22 | 3725 | |||
| NM_031387.3 | NP_113564.2 | 23 | 3585 | |||
| GenBank mRNA | AB067571.1 | - | 22 | 3406 | ||
| AB183526.1 | - | 23 | 3452 | |||
| AB183527.1 | - | 23 | 3425 | |||
| AB183528.1 | - | 22 | 3725 | |||
| AB183529.1 | - | 22 | 3289 | |||
| AF285591.1 | - | 19 | 2779 | |||
| AK029806.1 | - | 23 | 3586 | |||

Tdrd1
Genomic Map