siRNA Details
| Gene Symbol: Bicc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031397.2 | NP_113574.1 | 18 | 2543 | ||
| GenBank mRNA | AF319464.1 | - | 18 | 2544 | ||
| AK047271.1 | - | 8 | 1709 | |||
| AK076154.1 | - | 18 | 2596 | |||
| AK089066.1 | - | 18 | 2374 | |||
| BC062174.1 | - | 10 | 1412 | |||
| BC111523.2 | - | 18 | 2544 | 40048106 | ||

Bicc1
Genomic Map