siRNA Details
| Gene Symbol: Bicc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031397.2 | NP_113574.1 | 4, 5 | 421 | ||
| GenBank mRNA | AF319464.1 | - | 4, 5 | 422 | ||
| AK076154.1 | - | 4, 5 | 474 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bicc1| Gene Symbol: Bicc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031397.2 | NP_113574.1 | 4, 5 | 421 | ||
| GenBank mRNA | AF319464.1 | - | 4, 5 | 422 | ||
| AK076154.1 | - | 4, 5 | 474 | |||