siRNA Details
| Gene Symbol: Slc12a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031406.3 | NP_113583.2 | 7 | 1090 | ||
| GenBank mRNA | AF314957.1 | - | 6 | 956 | ||
| AK154118.1 | - | 7 | 1049 | |||
| BC046982.1 | - | 7 | 1010 | 6478046 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc12a9| Gene Symbol: Slc12a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031406.3 | NP_113583.2 | 7 | 1090 | ||
| GenBank mRNA | AF314957.1 | - | 6 | 956 | ||
| AK154118.1 | - | 7 | 1049 | |||
| BC046982.1 | - | 7 | 1010 | 6478046 | ||