siRNA Details
| Gene Symbol: Wasf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031877.3 | NP_114083.1 | 9 | 1735 | ||
| GenBank mRNA | AF290877.1 | - | 8 | 1631 | ||
| AF467773.1 | - | 8 | 1631 | |||
| BC016896.1 | - | 9 | 1693 | 4239038 | ||
| BC053423.1 | - | 9 | 1696 | 6509953 | ||

Wasf1
Genomic Map