siRNA Details
| Gene Symbol: Tas1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031873.1 | NP_114079.1 | 5, 6 | 1633 | ||
| GenBank mRNA | AF337041.1 | - | 5, 6 | 1633 | ||
| AY032623.1 | - | 5, 6 | 1602 | |||
| BC137740.1 | - | 6, 7 | 1535 | 8860759 | ||
| BC144879.1 | - | 6, 7 | 1535 | 9053427 | ||
More Information
Set Membership: |
|

Tas1r2
Genomic Map