siRNA Details
| Gene Symbol: Tmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033759.2 | NP_001028931.1 | 19 | 3740 | ||
| NM_031997.4 | NP_114386.3 | 20 | 3822 | |||
| GenBank mRNA | AF137031.1 | - | 2 | 185 | ||
| AK129352.1 | - | 14 | 2041 | |||
| BC019745.1 | - | 11 | 1463 | 3675225 | ||
| BC076570.1 | - | 19 | 3346 | 6830791 | ||
| BC089353.1 | - | 19 | 3722 | 6410437 | ||
| BG244335.1 | - | 438 | ||||
| BI666286.1 | - | 3 | 357 | |||

Tmem2
Genomic Map