siRNA Details
| Gene Symbol: Tmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033759.2 | NP_001028931.1 | 2 | 506 | ||
| NM_031997.4 | NP_114386.3 | 3 | 588 | |||
| GenBank mRNA | AK149667.1 | - | 3 | 561 | ||
| AK149803.1 | - | 3 | 543 | |||
| AK156343.1 | - | 2 | 459 | |||
| AK172599.1 | - | 2 | 506 | |||
| AK172691.1 | - | 2 | 468 | |||
| BC076570.1 | - | 2 | 112 | 6830791 | ||
| BC089353.1 | - | 2 | 488 | 6410437 | ||
| BU058152.1 | - | 2 | 506 | |||
| CA945463.1 | - | 3 | 444 | |||

Tmem2
Genomic Map