siRNA Details
| Gene Symbol: Tmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033759.2 | NP_001028931.1 | 10 | 2483 | ||
| NM_031997.4 | NP_114386.3 | 11 | 2565 | |||
| GenBank mRNA | AK129352.1 | - | 5 | 784 | ||
| BC019745.1 | - | 2 | 206 | 3675225 | ||
| BC076570.1 | - | 10 | 2089 | 6830791 | ||
| BC089353.1 | - | 10 | 2465 | 6410437 | ||
| CA752446.1 | - | 3 | 423 | |||

Tmem2
Genomic Map