siRNA Details
| Gene Symbol: Cep41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ278891.1 | - | 10 | 978 | ||
| AK006059.1 | - | 10 | 966 | |||
| AK132495.1 | - | 10 | 967 | |||
| AK161461.1 | - | 10 | 952 | |||
| AK166974.1 | - | 11 | 1095 | |||
| AK170213.1 | - | 10 | 991 | |||
| BC031892.1 | - | 10 | 948 | 5365552 | ||

Cep41
Genomic Map