siRNA Details
| Gene Symbol: Btbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146193.2 | NP_666305.2 | 5 | 1058 | ||
| GenBank mRNA | AK088345.1 | - | 5 | 1099 | ||
| BC031192.1 | - | 4 | 536 | 5043252 | ||
| BC072618.1 | - | 5 | 1058 | 6811133 | ||
| BC087937.1 | - | 5 | 1091 | 6705214 | ||

Btbd1
Genomic Map