siRNA Details
| Gene Symbol: Tbx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032005.4 | NP_114394.1 | 2 | 515 | ||
| GenBank mRNA | AF345520.1 | - | 2 | 341 | ||
| AF348321.1 | - | 2 | 472 | |||
| AK038666.1 | - | 2 | 517 | |||
| BC120635.1 | - | 2 | 386 | 40129558 | ||
| BC137731.1 | - | 2 | 386 | 8860750 | ||

Tbx19
Genomic Map