siRNA Details
| Gene Symbol: Mmp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032007.3 | NP_114396.3 | 4 | 612 | ||
| GenBank mRNA | AJ278461.1 | - | 4 | 581 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp1b| Gene Symbol: Mmp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032007.3 | NP_114396.3 | 4 | 612 | ||
| GenBank mRNA | AJ278461.1 | - | 4 | 581 | ||