siRNA Details
| Gene Symbol: Slmap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032008.3 | NP_114397.3 | 7 | 854 | ||
| GenBank mRNA | AK052158.1 | - | 8 | 1820 | ||
| AK129403.1 | - | 7 | 854 | |||
| AK141597.1 | - | 7 | 2011 | |||
| AK163630.1 | - | 8 | 1793 | |||
| AK164911.1 | - | 7 | 2009 | |||
| AK166396.1 | - | 7 | 2010 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106060, XM_008770981, XM_006252544, XM_006252543, XM_006252550, and others.

Slmap
Genomic Map