siRNA Details
| Gene Symbol: Plvap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032398.2 | NP_115774.2 | 1, 2 | 427 | ||
| GenBank mRNA | AF348403.1 | - | 1, 2 | 398 | ||
| AF369900.1 | - | 1, 2 | 422 | |||
| BC013517.1 | - | 1, 2 | 430 | 4235957 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plvap| Gene Symbol: Plvap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032398.2 | NP_115774.2 | 1, 2 | 427 | ||
| GenBank mRNA | AF348403.1 | - | 1, 2 | 398 | ||
| AF369900.1 | - | 1, 2 | 422 | |||
| BC013517.1 | - | 1, 2 | 430 | 4235957 | ||