siRNA Details
| Gene Symbol: Hamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032541.1 | NP_115930.1 | 3 | 246 | ||
| GenBank mRNA | AF297664.1 | - | 3 | 246 | ||
| AF503444.1 | - | 3 | 246 | |||
| BC021587.1 | - | 3 | 229 | 5097807 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hamp| Gene Symbol: Hamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032541.1 | NP_115930.1 | 3 | 246 | ||
| GenBank mRNA | AF297664.1 | - | 3 | 246 | ||
| AF503444.1 | - | 3 | 246 | |||
| BC021587.1 | - | 3 | 229 | 5097807 | ||