siRNA Details
| Gene Symbol: Sval2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032542.1 | NP_115931.1 | 2 | 111 | ||
| GenBank mRNA | AB050614.1 | - | 2 | 111 | ||
| BC119343.1 | - | 2 | 211 | 8734092 | ||
| BC119345.1 | - | 2 | 211 | 8734094 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sval2| Gene Symbol: Sval2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032542.1 | NP_115931.1 | 2 | 111 | ||
| GenBank mRNA | AB050614.1 | - | 2 | 111 | ||
| BC119343.1 | - | 2 | 211 | 8734092 | ||
| BC119345.1 | - | 2 | 211 | 8734094 | ||