siRNA Details
| Gene Symbol: Cd96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032465.2 | NP_115854.2 | 6, 7 | 1042 | ||
| GenBank mRNA | AK040329.1 | - | 6, 7 | 929 | ||
| AK040539.1 | - | 6, 7 | 1046 | |||
| AK154076.1 | - | 6, 7 | 1012 | |||
| AK156470.1 | - | 6, 7 | 1043 | |||
| AY029156.1 | - | 6, 7 | 903 | |||
| BC052865.1 | - | 6, 7 | 939 | 30072864 | ||

Cd96
Genomic Map