siRNA Details
| Gene Symbol: Rnf123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032543.2 | NP_115932.1 | 27 | 2714 | ||
| GenBank mRNA | AK137599.1 | - | 26 | 3064 | ||
| AK157870.1 | - | 27 | 2642 | |||
| AK172244.1 | - | 27 | 2639 | |||
| AY744153.1 | - | 26 | 2513 | |||
| BC025530.1 | - | 27 | 2635 | |||
| BC030482.1 | - | 2 | 68 | |||
| BC057082.1 | - | 27 | 3127 | 5704926 | ||

Rnf123
Genomic Map