siRNA Details
| Gene Symbol: Rnf123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032543.2 | NP_115932.1 | 32, 33 | 3346 | ||
| GenBank mRNA | AK137599.1 | - | 31, 32 | 3696 | ||
| AK157870.1 | - | 32, 33 | 3274 | |||
| AK172244.1 | - | 32, 33 | 3271 | |||
| AY744153.1 | - | 31, 32 | 3145 | |||
| BC003945.1 | - | 6, 7 | 551 | |||
| BC025530.1 | - | 32, 33 | 3267 | |||
| BC030482.1 | - | 7, 8 | 700 | |||
| BC057082.1 | - | 32, 33 | 3759 | 5704926 | ||

Rnf123
Genomic Map