siRNA Details
| Gene Symbol: Rnf123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032543.2 | NP_115932.1 | 36 | 3755 | ||
| GenBank mRNA | AK137599.1 | - | 35 | 4105 | ||
| AK172244.1 | - | 36 | 3680 | |||
| AK212142.1 | - | 3 | 195 | |||
| AK216931.1 | - | 4 | 377 | |||
| AY744153.1 | - | 35 | 3554 | |||
| BC003945.1 | - | 10 | 976 | |||
| BC025530.1 | - | 36 | 3676 | |||
| BC030482.1 | - | 11 | 1109 | |||
| BC057082.1 | - | 36 | 4168 | 5704926 | ||

Rnf123
Genomic Map