siRNA Details
| Gene Symbol: Kars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130868.2 | NP_001124340.1 | 5, 6 | 745 | ||
| NM_001286384.1 | NP_001273313.1 | 6, 7 | 803 | |||
| NM_053092.3 | NP_444322.1 | 4, 5 | 565 | |||
| GenBank mRNA | AK089053.1 | - | 4, 5 | 490 | ||
| AK090203.1 | - | 4, 5 | 490 | |||
| AK153236.1 | - | 4, 5 | 488 | |||
| AK167693.1 | - | 4, 5 | 490 | |||
| BC027356.1 | - | 6, 7 | 715 | 4950333 | ||
| BC035324.1 | - | 5, 6 | 657 | 2811286 | ||
| BC036289.1 | - | 4, 5 | 534 | 3157706 | ||

Kars
Genomic Map