siRNA Details
| Gene Symbol: Tac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053093.1 | NP_444323.1 | 2 | 316 | ||
| GenBank mRNA | AF235035.1 | - | 2 | 316 | ||
| AF515827.1 | - | 2 | 150 | |||
| BC119426.1 | - | 2 | 296 | 40039772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tac4| Gene Symbol: Tac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053093.1 | NP_444323.1 | 2 | 316 | ||
| GenBank mRNA | AF235035.1 | - | 2 | 316 | ||
| AF515827.1 | - | 2 | 150 | |||
| BC119426.1 | - | 2 | 296 | 40039772 | ||