siRNA Details
| Gene Symbol: Cd163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170395.1 | NP_001163866.1 | 6 | 1364 | ||
| NM_053094.2 | NP_444324.2 | 6 | 1364 | |||
| GenBank mRNA | AF274883.1 | - | 6 | 1359 | ||
| BC144848.1 | - | 6 | 1315 | 9053396 | ||
| BC145793.1 | - | 6 | 1315 | 40131096 | ||
| DQ058616.1 | - | 6 | 1320 | |||
| DQ058617.1 | - | 6 | 1320 | |||

Cd163
Genomic Map