siRNA Details
| Gene Symbol: Trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053100.2 | NP_444330.2 | 3 | 1498 | ||
| GenBank mRNA | AF220035.1 | - | 2 | 117 | ||
| AF281047.1 | - | 3 | 847 | |||
| AK052763.1 | - | 3 | 233 | |||
| AK155303.1 | - | 3 | 1445 | |||
| BC029283.1 | - | 3 | 1432 | |||
| BC037065.1 | - | 3 | 783 | |||
| BC060094.1 | - | 3 | 728 | |||
| BC082298.1 | - | 3 | 1373 | |||
| BC152343.1 | - | 3 | 834 | 40093266 | ||
| BC152344.1 | - | 3 | 834 | 40093269 | ||

Trim8
Genomic Map