siRNA Details
| Gene Symbol: Trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053100.2 | NP_444330.2 | 6 | 2214 | ||
| GenBank mRNA | AF281047.1 | - | 6 | 1563 | ||
| AK052763.1 | - | 6 | 949 | |||
| AK079847.1 | - | 4 | 653 | |||
| AK155303.1 | - | 6 | 2161 | |||
| BC060094.1 | - | 6 | 1444 | |||
| BC082298.1 | - | 6 | 2089 | |||
| BC152343.1 | - | 6 | 1550 | 40093266 | ||
| BC152344.1 | - | 6 | 1550 | 40093269 | ||

Trim8
Genomic Map