siRNA Details
| Gene Symbol: Trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053100.2 | NP_444330.2 | 1, 2 | 1358 | ||
| GenBank mRNA | AF281047.1 | - | 1, 2 | 707 | ||
| AK052763.1 | - | 1, 2 | 93 | |||
| AK155303.1 | - | 1, 2 | 1305 | |||
| BC029283.1 | - | 1, 2 | 1292 | |||
| BC037065.1 | - | 1, 2 | 643 | |||
| BC060094.1 | - | 1, 2 | 588 | |||
| BC082298.1 | - | 1, 2 | 1233 | |||
| BC152343.1 | - | 1, 2 | 694 | 40093266 | ||
| BC152344.1 | - | 1, 2 | 694 | 40093269 | ||

Trim8
Genomic Map