siRNA Details
| Gene Symbol: Zkscan8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251833.1 | NP_001238762.1 | 4 | 1398 | ||
| NM_139141.4 | NP_631880.2 | 6 | 1614 | |||
| GenBank mRNA | AK032718.1 | - | 6 | 1570 | ||
| AK134299.1 | - | 6 | 1599 | |||
| BC050152.1 | - | 6 | 1550 | |||
| BC145448.1 | - | 3 | 1232 | 9054006 | ||
| BC150775.1 | - | 3 | 1232 | 9087686 | ||

Zkscan8
Genomic Map