siRNA Details
| Gene Symbol: Klhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053105.2 | NP_444335.2 | 4 | 1665 | ||
| GenBank mRNA | AF252281.1 | - | 4 | 1594 | ||
| AK031932.1 | - | 4 | 1477 | |||
| AK220397.1 | - | 4 | 969 | |||
| BC094584.1 | - | 4 | 1665 | 30547502 | ||

Klhl1
Genomic Map