siRNA Details
Gene Symbol: Gpr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053107.4 | NP_444337.3 | 2 | 940 | ||
GenBank mRNA | AK039769.1 | - | 2 | 850 | ||
BC118506.1 | - | 1 | 470 | 40106287 | ||
BC118510.1 | - | 1 | 470 | 40106284 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
