siRNA Details
| Gene Symbol: Pcdhgb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033578.3 | NP_291056.1 | 1 | 2381 | ||
| GenBank mRNA | AY013808.1 | - | 1 | 2234 | ||
| BC054556.1 | - | 1 | 2381 | 5702016 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhgb6| Gene Symbol: Pcdhgb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033578.3 | NP_291056.1 | 1 | 2381 | ||
| GenBank mRNA | AY013808.1 | - | 1 | 2234 | ||
| BC054556.1 | - | 1 | 2381 | 5702016 | ||