siRNA Details
| Gene Symbol: Pabpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053114.2 | NP_444344.1 | 2 | 474 | ||
| GenBank mRNA | AK050551.1 | - | 2 | 476 | ||
| BC107362.1 | - | 1 | 375 | 40056131 | ||
| BC107363.2 | - | 1 | 375 | 40056132 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pabpc5| Gene Symbol: Pabpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053114.2 | NP_444344.1 | 2 | 474 | ||
| GenBank mRNA | AK050551.1 | - | 2 | 476 | ||
| BC107362.1 | - | 1 | 375 | 40056131 | ||
| BC107363.2 | - | 1 | 375 | 40056132 | ||