siRNA Details
| Gene Symbol: Sirt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159589.1 | NP_001153061.1 | 8 | 2028 | ||
| NM_019812.2 | NP_062786.1 | 9 | 2142 | |||
| GenBank mRNA | AF214646.1 | - | 9 | 2127 | ||
| AK139946.1 | - | 2 | 637 | |||
| AK144203.1 | - | 8 | 2031 | |||
| AK213131.1 | - | 1 | 119 | |||
| AY377984.1 | - | 9 | 2127 | |||
| BC006584.1 | - | 2 | 448 | |||
| BC152314.1 | - | 9 | 1622 | 40111573 | ||

Sirt1
Genomic Map