siRNA Details
| Gene Symbol: Sirt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159589.1 | NP_001153061.1 | 4 | 942 | ||
| NM_019812.2 | NP_062786.1 | 5 | 1056 | |||
| GenBank mRNA | AF214646.1 | - | 5 | 1041 | ||
| AK144203.1 | - | 4 | 945 | |||
| AY377984.1 | - | 5 | 1041 | |||
| BC152314.1 | - | 5 | 536 | 40111573 | ||

Sirt1
Genomic Map