siRNA Details
| Gene Symbol: Smarca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053124.2 | NP_444354.2 | 17 | 2624 | ||
| GenBank mRNA | AF325921.1 | - | 17 | 2276 | ||
| AF375046.1 | - | 17 | 2343 | |||
| AK052320.1 | - | 5 | 577 | |||
| AK153789.1 | - | 17 | 2942 | |||
| AK156915.1 | - | 1 | 88 | |||
| AK157142.1 | - | 2635 | ||||
| BC021922.1 | - | 5 | 562 | |||
| BC053069.1 | - | 17 | 2624 | 6409140 | ||

Smarca5
Genomic Map