siRNA Details
| Gene Symbol: Peli2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033602.2 | NP_291080.2 | 5 | 817 | ||
| GenBank mRNA | AF302504.1 | - | 5 | 746 | ||
| AK030564.1 | - | 5 | 864 | |||
| AK033815.1 | - | 2 | 123 | |||
| AK045515.1 | - | 4 | 690 | |||
| BC027062.1 | - | 4 | 656 | 5355240 | ||

Peli2
Genomic Map