siRNA Details
| Gene Symbol: Peli2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033602.2 | NP_291080.2 | 2 | 377 | ||
| GenBank mRNA | AF302504.1 | - | 2 | 306 | ||
| AK030564.1 | - | 2 | 424 | |||
| AK082342.1 | - | 2 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Peli2| Gene Symbol: Peli2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033602.2 | NP_291080.2 | 2 | 377 | ||
| GenBank mRNA | AF302504.1 | - | 2 | 306 | ||
| AK030564.1 | - | 2 | 424 | |||
| AK082342.1 | - | 2 | 333 | |||