siRNA Details
| Gene Symbol: Dqx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033606.3 | NP_291084.1 | 11 | 2213 | ||
| GenBank mRNA | AF318278.1 | - | 10 | 1857 | ||
| BC048936.1 | - | 6 | 876 | |||
| BC075680.1 | - | 11 | 2213 | 30612145 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dqx1| Gene Symbol: Dqx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033606.3 | NP_291084.1 | 11 | 2213 | ||
| GenBank mRNA | AF318278.1 | - | 10 | 1857 | ||
| BC048936.1 | - | 6 | 876 | |||
| BC075680.1 | - | 11 | 2213 | 30612145 | ||