siRNA Details
| Gene Symbol: Uchl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033607.1 | NP_291085.1 | 1 | 153 | ||
| GenBank mRNA | AB035420.1 | - | 1 | 153 | ||
| BC139440.1 | - | 1 | 144 | 8862462 | ||
| BC139441.1 | - | 1 | 144 | 8862463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uchl4| Gene Symbol: Uchl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033607.1 | NP_291085.1 | 1 | 153 | ||
| GenBank mRNA | AB035420.1 | - | 1 | 153 | ||
| BC139440.1 | - | 1 | 144 | 8862462 | ||
| BC139441.1 | - | 1 | 144 | 8862463 | ||