siRNA Details
| Gene Symbol: Igsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145800.1 | NP_001139272.1 | 20 | 3519 | ||
| NM_033608.3 | NP_291086.2 | 20 | 3585 | |||
| GenBank mRNA | AF317839.1 | - | 20 | 3585 | ||
| AK078536.1 | - | 20 | 3515 | |||
| AK145121.1 | - | 20 | 3499 | |||
| BC023011.1 | - | 2 | 379 | |||
| BC024487.1 | - | 2 | 357 | |||
| BC034594.1 | - | 19 | 3348 | |||
| BC040281.1 | - | 20 | 3545 | 5364073 | ||

Igsf9
Genomic Map