siRNA Details
| Gene Symbol: Pnck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199351.1 | NP_001186280.1 | 5 | 484 | ||
| NM_001199352.1 | NP_001186281.1 | 5 | 348 | |||
| NM_012040.3 | NP_036170.1 | 5 | 424 | |||
| GenBank mRNA | AB023027.1 | - | 4 | 294 | ||
| AF181984.1 | - | 5 | 398 | |||
| AK134665.1 | - | 5 | 355 | |||
| AK158755.1 | - | 5 | 349 | |||
| BC051996.1 | - | 5 | 331 | |||
| BC055891.1 | - | 5 | 324 | 4208888 | ||
| CX243772.1 | - | 5 | 484 | |||

Pnck
Genomic Map