siRNA Details
| Gene Symbol: Pnck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199351.1 | NP_001186280.1 | 8 | 837 | ||
| NM_001199352.1 | NP_001186281.1 | 8 | 701 | |||
| NM_012040.3 | NP_036170.1 | 8 | 777 | |||
| GenBank mRNA | AB023027.1 | - | 7 | 647 | ||
| AF181984.1 | - | 8 | 751 | |||
| AK134665.1 | - | 8 | 708 | |||
| AK158755.1 | - | 8 | 702 | |||
| BC051996.1 | - | 8 | 684 | |||
| BC055891.1 | - | 8 | 677 | 4208888 | ||

Pnck
Genomic Map