siRNA Details
| Gene Symbol: Pcdhb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053128.2 | NP_444358.2 | 1 | 1381 | ||
| GenBank mRNA | AF326296.1 | - | 1 | 1197 | ||
| AK030570.1 | - | 1 | 1381 | |||
| AY013785.1 | - | 1 | 1197 | |||
| BC079557.1 | - | 1 | 1295 | 30360109 | ||

Pcdhb3
Genomic Map