siRNA Details
| Gene Symbol: Pcdhb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053129.3 | NP_444359.3 | 1 | 872 | ||
| GenBank mRNA | AK029277.1 | - | 1 | 864 | ||
| AK036169.1 | - | 1 | 873 | |||
| AK143280.1 | - | 1 | 873 | |||
| AK160730.1 | - | 1 | 873 | |||
| AY013786.1 | - | 1 | 688 | |||
| BC053098.1 | - | 1 | 845 | 6401988 | ||

Pcdhb4
Genomic Map