siRNA Details
| Gene Symbol: Pcdhb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053131.1 | NP_444361.1 | 1 | 685 | ||
| GenBank mRNA | AK032193.1 | - | 298 | |||
| AY013788.1 | - | 1 | 685 | |||
| BC150716.1 | - | 1 | 767 | 9087627 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhb6| Gene Symbol: Pcdhb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053131.1 | NP_444361.1 | 1 | 685 | ||
| GenBank mRNA | AK032193.1 | - | 298 | |||
| AY013788.1 | - | 1 | 685 | |||
| BC150716.1 | - | 1 | 767 | 9087627 | ||