siRNA Details
| Gene Symbol: Pcdhb22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053147.3 | NP_444377.3 | 1 | 375 | ||
| GenBank mRNA | AF326315.1 | - | 1 | 247 | ||
| AK043492.1 | - | 1 | 376 | |||
| AK143176.1 | - | 1 | 856 | |||
| AY013784.1 | - | 1 | 247 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhb22| Gene Symbol: Pcdhb22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053147.3 | NP_444377.3 | 1 | 375 | ||
| GenBank mRNA | AF326315.1 | - | 1 | 247 | ||
| AK043492.1 | - | 1 | 376 | |||
| AK143176.1 | - | 1 | 856 | |||
| AY013784.1 | - | 1 | 247 | |||