siRNA Details
| Gene Symbol: Nkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163660.1 | NP_001157132.1 | 5 | 431 | ||
| NM_027280.3 | NP_081556.3 | 7 | 641 | |||
| GenBank mRNA | AF343352.1 | - | 7 | 478 | ||
| AF358134.1 | - | 7 | 557 | |||
| AK076164.1 | - | 7 | 632 | |||
| AK082367.1 | - | 7 | 630 | |||
| AK166564.1 | - | 7 | 644 | |||
| BC034838.1 | - | 7 | 615 | 1429943 | ||
| DV662736.1 | - | 5 | 411 | |||

Nkd1
Genomic Map